Learn More
MilliporeSigma™ Mayer's Hemalum Solutions for Microscopy
Catalog No. M1092490500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500mL
1L
2.5L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1092490500 500mL
M1092491000 1L
M1092492500 2.5L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1092490500 Supplier MilliporeSigma™ Supplier No. 1.09249.0500
Add to Cart
Add to Cart
for microscopy
In hematoxylin-eosin staining (HandE), the most widely used histological staining technique, nuclear staining is achieved using ready-to-use staining solutions, such as Mayer’s hemalum solution or Hematoxylin solution modified according to Gill III. These solutions do not need to be filtrated prior use and the dye is oxidized in a controlled manner, which renders the results robust and allows a brilliant differentiation of the stained structures. Counterstaining of i.e. for proteins, collagen, keratin, intercellular substances. can be performed with alcoholic or aqueous Eosin Y solution 0.5 %. It is registered as IVD and CE certified product and can be used for clinical diagnostics.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 500mL
For Use With (Application) Microscopy
Product Type Hemalum Solution