Learn More
MilliporeSigma™ MF2 Push-button Manifold
Catalog No. 6015 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1/Pk.
Catalog No. Quantity
6015 1/Pk.
1 options
Catalog No. 6015 Supplier MilliporeSigma™ Supplier No. 6015
Add to Cart
Add to Cart
For instant direction of gas pressure or liquid flow in multi-cell or multi-reservoir systems
  • Can pressurize up to three cells or reservoirs from one gas source or feed several cells from one reservoir
  • Several manifolds can be combined to operate more than three units in parallel
  • Made from delrin plastic, Buna-N material
  • Chemically sanitizable
  • Dimensions: 4 x 2 x 2 in. (10 x 5 x 5cm)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Allows for operation of multiple cells or reservoirs
Material Delrin plastic, Buna-N
Quantity 1/Pk.