Learn More
MilliporeSigma™ Microcon DNA Fast Flow PCR Grade with Ultracel Membrane, Dual Cycle EtO Treated
Shop All MilliporeSigma ProductsDescription
Microcon PCR Grade Centrifugal Filters are ideal for DNA concentration and desalting applications. The dual cycle Ethylene Oxide (EtO) treatment renders contaminating DNA non-amplifiable. Available in a pack of 20.
Specifications
Specifications
| Color | Blue |
| Disposable | Single-use |
| Material (Membrane) | Regenerated Cellulose |
| MWCO | 100 kDa |
| Sterility | Non-sterile |
| Filtration Area | 0.32 cm2 |
| For Use With (Application) | DNA Concentration and Desalting |
| Housing Material | Regenerated Cellulose (RC) |
| Membrane Type | Ultracell low binding regenerated cellulose |
| Quantity | 0.5 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.