Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Microcon DNA Fast Flow Centrifugal Filter Unit with Ultracel Membrane
GSA_VA

Catalog No. MRCF0R100 Shop All MilliporeSigma Products
Change view
Click to view available options
MWCO:
100 kDa
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO
MRCF0R100 100 kDa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MRCF0R100 Supplier MilliporeSigma™ Supplier No. MRCF0R100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

This 0.5 mL centrifugal filter is optimized for the concentration and recovery of genomic DNA with SDS buffer.

Optimized for the concentration and recovery of genomic DNA.

Simply and efficiently concentrate and desalt solutions of any macromolecule, using any centrifuge that can accept 1.5 mL tubes. With the low-binding Ultracel™ membrane, Microcon™ filters offer:•Typical recoveries of greater than 95%, even for dilute solutions •Reverse spin to maximize recovery, even in the smallest samples •Convenient storage of filtrate or concentrated sample in standard microfuge tube •Concentration factors up to 100X Microcon™ DNA Fast Flow FilterOptimized for the concentration and recovery of genomic DNA with SDS buffer. The low nonspecific binding characteristics of the membrane and the other device components, coupled with its medical-grade o-ring seal, allows the device to accommodate several wash steps with minimal sample loss.Features:•High recovery for small volumes with reverse spin (concentration factor less than 20X) •Low-binding Ultracel™ membrane •Fast processing Sample applications:•Recovery of genomic DNA for forensic applications •Concentration and desalting of nucleic acids (high recovery alternative to ethanol precipitation) •Removal of primers from amplified DNA •Easy filtrate recovery, exploiting the flat membrane design

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Disposable Single-use
Material (Membrane) Regenerated Cellulose
MWCO 100 kDa
Sterility Non-sterile
Filtration Area 0.32 cm2
Housing Material Regenerated Cellulose (RC)
Type Centrifugal Filters
Volume (Metric) 10 mL
Length (English) 1.8 in.
Length (Metric) 4.5 cm
Diameter (Metric) 12.3 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.