Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Microscope Slides

Catalog No. XX1007615 Shop All MilliporeSigma Products
Change view
Click to view available options
2 x 3 in. dimensions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
XX1007615 2 x 3 in. dimensions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. XX1007615 Supplier MilliporeSigma™ Supplier No. XX1007615
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Accessories for filter holders for sample preparation.

Accessories for filter holders for sample preparation.
TRUSTED_SUSTAINABILITY

Specifications

Sterility Non-sterile
No. per Pack 72
Color Clear
Coating Uncoated
For Use With (Application) Accessories for filter holders for sample preparation
Length (English) 2 in.
Width (English) 2 in.
Dimensions (L x W) 2 x 3 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.