Learn More
MilliporeSigma™ MIDAS™ III Automated Slide Stainer Accessory, Vessel and Lid
Catalog No. M64054W95 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Product Type:
Vessel and Lid
Catalog No. Product Type
M64054W95 Vessel and Lid
1 options
Catalog No. M64054W95 Supplier MilliporeSigma™ Supplier No. 64054W95
Add to Cart
Add to Cart
For use with MIDAS™ III Automated Stainer
  • Snug fitting lids to protect the stains and solutions from evaporation and contamination while not in use.
  • Inexpensive, disposable staining vessels eliminate cleaning of jars
  • See-thru fill marks 50mL, 75mL, and 300mL
  • Includes 8 vessels and 8 lids
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vessel and Lid
For Use With (Equipment) MIDAS™ III Automated Slide Stainer