Learn More
MilliporeSigma™ MIDAS™ III Automated Slide Stainer Accessory, Ryton Vessel and Lid
Shop All MilliporeSigma ProductsDescription
- Snug fitting lids to protect the stains and solutions from evaporation and contamination while not in use.
- Inexpensive, disposable staining vessels eliminate cleaning of jars
- Includes 2 vessels and 2 lids
Specifications
Specifications
| Product Type | Ryton Vessels and Lids |
| For Use With (Equipment) | MIDAS™ III Automated Slide Stainer |