Learn More
MilliporeSigma™ MIDAS™ III Automated Slide Stainer Accessory, Staining Rack System
Shop All MilliporeSigma ProductsDescription
- Snug fitting lids to protect the stains and solutions from evaporation and contamination while not in use.
- Solvent resistant drip tray to keep messy stains off your countertops.
- Inexpensive, disposable staining vessels eliminate cleaning of jars
- Additional components are sold separately to allow you to change to other Staining procedures or replenish your stains solutions easily and quickly.
Specifications
Specifications
| Product Type | Staining Rack System |
| For Use With (Equipment) | MIDAS™ III Automated Slide Stainer |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.