Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Midas™ Slide Carrier

Catalog No. M64052E94 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Product Type:
Slide Carrier
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
M64052E94 Slide Carrier
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M64052E94 Supplier MilliporeSigma™ Supplier No. 64052E94
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Holds up to 30 slides in MIDAS™ Automated Stainer.

  • Holds 30 slides of size 7.62 × 2.54cm (3 × 1 in.)
  • Store at room temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type Slide Carrier
Slide Capacity 30
For Use With (Equipment) MIDAS Automated Stainer
Quantity 1 Slide Carrier
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.