Learn More
MilliporeSigma™ Replacement Electrodes for Millicell-ERS Volt-Ohm Meter, 1 pair
Catalog No. MERSSTX01 Shop All MilliporeSigma Products
Change view
Click to view available options
Replacement Electrodes
Catalog No.
MERSSTX01 Replacement Electrodes
1 options
Catalog No. MERSSTX01 Supplier MilliporeSigma™ Supplier No. MERSSTX01
Measures Trans Epithelial Electrical Resistance (TEER) of epithelial cells in culture
Provides qualitative measurement of cell monolayer health and quantative measures of confluency. Designed for use with cells cultured in Millicell™ Culture Plate Inserts only.
Utilizes AC current for membrane resistance measurements, producing a zero net charge on cells and eliminating adverse DC current effects on cell membranes. A silver/silver chloride (Ag/AgCI) pellet on each electrode tip measures voltage. Due to the small size of the electrodes, the user can easily measure transepithelial voltage and the resistance of cells grown on microporous membranes.
Warranty and Services
Electrodes and batteries are warranted for 30 days only from the date on which the customer receives these items
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Millicell-ERS Volt-Ohm Meter
For Use With (Application) To provides qualitative measurement of cell monolayer health and quantative measures of confluency
Product Type Replacement Electrode
Frequency 12.5 Hz