Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MMP-2, Active, Human, Recombinant Protein, Mouse Cells

Catalog No. PF0235UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PF0235UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PF0235UG Supplier MilliporeSigma™ Supplier No. PF0235UG
Only null left
Add to Cart
Add to Cart
Full-length, recombinant, human pro-MMP-2 expressed in Mouse Cells that is subsequently activated by APMA. APMA is removed through a desalting column. The substrate specificity for MMP-2 includes collagen (types IV, V, VII, and X), elastin, and gelatin (type I). The presence of TIMP-2 inhibitor prevents degradation of the MMP-2 C-terminal regulatory domain. TIMP-2 is also an inhibitor of proteolysis and will inhibit the activity of the enzyme. Useful for immunoblotting, substrate cleavage assay and zymography.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 50mM HEPES, 10mM CaCl2, 20% glycerol, 0.005% BRIJ™-35 Detergent, pH 7.5.
Purification Method SDS-PAGE; contains 9% TIMP-2
Quantity 5 μg
Immunogen MMP-2
Storage Requirements Following initial use, aliquot into siliconized vials and freeze (−70°C).
Regulatory Status RUO
Cell Type Mouse Cells
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.