Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MMP-9, Dimer, Human Neutrophil, Protein

Catalog No. 4442325UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-423-25UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-423-25UG Supplier MilliporeSigma™ Supplier No. 4442325UG
Only null left
Add to Cart
Add to Cart
Dimeric, native MMP-9 from stimulated human neutrophils. Enzyme should be activated just prior to use.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 200mM NaCl, 50mM Tris-HCl, 5mM CaCl2, 1μM ZnCl2, 0.05% BRIJ™ 35 Detergent, 0.05% NaN3, pH 7.0.
Purification Method SDS-PAGE
Quantity 5 μg
Immunogen MMP-9
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Structural Form Dimer
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.