Learn More
MilliporeSigma™ MMP-9-Lipocalin-TIMP-1 Complex, Human Neutrophil, Protein
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Formulation | In 200mM NaCl, 50mM Tris-HCl, 5mM CaCl2, 1μM ZnCl2, 0.05% BRIJ™ 35 Detergent, 0.05% NaN3, pH 7.0. |
| Purification Method | SDS-PAGE |
| Quantity | 5 μg |
| Immunogen | MMP-9-Lipocalin-TIMP-1 Complex |
| Storage Requirements | Following initial thaw, aliquot and freeze (−70°C). |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Native |
| Form | Liquid |
| Purity or Quality Grade | ≥90% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.