Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MMP/TIMP Family Sampler kit

Catalog No. ASK101EA Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ASK101EA 1 Kit Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ASK101EA Supplier MilliporeSigma™ Supplier No. ASK101EA
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects MMP/TIMP Family Clone: in samples, and it is validated for

Matrix metalloproteinases (MMPs) are a family of enzymes that are responsible for the degradation of extracellular matrix components such as collagen, laminin and proteoglycans. All MMPs share: sequence homology, a catalytic mechanism dependent upon a zinc ion at the active center, ability to cleave one or more extracellular matrix components, secretion as zymogens which are activated by removal of an approximately 10kDa segment from the N-terminus, and regulation by endogenous inhibitors. These enzymes are involved in normal physiological processes as well as in metastasis. MMP-2 and MMP-9 are members of the gelatinase family and elevated levels may be correlated with metastatic potential. MMP-1 is also known as interstitial collagenase and MMP-3 as stromelysin-1. Activation and activity of the MMP's are regulated by the TIMP (tissue inhibitors of metalloproteinase) family members. TIMP-1 preferentially regulates MMP-9 and TIMP-2 regulates MMP-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP/TIMP Family
Classification Monoclonal
Conjugate Unconjugated
Formulation 20μg of each antibody in 100mM sodium phosphate buffer, 0.1% BSA, pH 7.4.
Host Species Mouse
Quantity 1 Kit
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Contains 20μg of each antibody. Includes Anti-MMP-1, Anti-MMP-2, Anti-MMP-3, Anti-MMP-9, Anti-TIMP-1, and Anti-TIMP-2.
Includes 20μg of each antibody in 100mM sodium phosphate buffer, 0.1% BSA, pH 7.4.
Content And Storage −20°C
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.