Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Monomethyl-Histone H3 (Lys36), Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710498MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-104-98MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-104-98MI Supplier MilliporeSigma™ Supplier No. 1710498
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Specifically detects Monomethyl-Histone H3 (Lys36) Clone: in Human, Mouse, Rat samples, and it is validated for Chromatin Immunoprecipitation, Dot Blot, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Monomethyl-Histone H3 (Lys36)
Applications ChIP Assay, Dot Blot, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-Monomethyl-Histone H3 (Lys36) (Rabbit Polyclonal). One vial containing 75μL 0.35mg/mL purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl, and 0.05% sodium azide, before the addition of 30% glycerol. Normal Rabbit IgG. One vial containing 125μg Rabbit IgG in 125μL storage buffer containing 0.05% sodium azide. ChIP Primers, GAPDH coding D2. One vial containing 75μL of 5μM of each primer specific for human GAPDH coding region. FOR: GCC ATG TAG ACC CCT TGA AGA G; REV: ACT GGT TGA GCA CAG GGT ACT TTA T
Gene Accession No. Q16695
Gene Symbols HIST3H3, H3FT, H3/t, H3t, H3/g, H3.4, H3T
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the N-terminus of human Histone H3 monomethylated at Lys36.
Purification Method Affinity Purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_003484
Test Specificity This antibody recognizes the N-terminus of Histone H3 monomethylated at Lys36.
Includes The ChIPAb+ Monomethyl-Histone H3 (Lys36) set includes the monomethyl-Histone H3 (Lys36) antibody, a Normal Rabbit IgG and control primers which amplify a 213bp region of the human GAPDH coding region.
Target Species Human, Mouse, Rat
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.