Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Monomethyl-Histone H3 (Lys4)α, Mouse monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 17676MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-676-MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-676-MI Supplier MilliporeSigma™ Supplier No. 17676
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Specifically detects Monomethyl-Histone H3 (Lys4)α Clone: in Vertebrate samples, and it is validated for Chromatin Immunoprecipitation, Western Blotting

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the 'beads on a string' structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Monomethyl-Histone H3 (Lys4)α
Applications ChIP Assay, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-Monomethyl-HisPtone H3 (Lys4) (mouse monoclonal), One vial containing 50μg of protein G purified antibody in 50μL PBS containing 0.05% sodium azide. Normal Mouse IgG. Two vials containing 25μg purified Mouse IgG in 25μL storage buffer containing 0.1% sodium azide. ChIP Primers, human GAPDH coding region, . One vial containing 75μL of 5μM each primer specific for human GAPDH coding region. FOR: GGC TCC CAC CTT TCT CAT CC; REV: GGC CAT CCA CAG TCT TCT GG
Gene Accession No. Q16695
Gene Symbols HIST3H3; H3FT; H3/t; H3t; MGC126888; H3/g; H3.4; H3T; MGC126886
Host Species Mouse
Immunogen Peptide corresponding to Histone H3 containing the sequence [ART(me1-K)QTARKSTGC] on which Lys4 is acetylated.
Purification Method Protein A purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NM_003493
Test Specificity This antibody detects Monomethyl-Histone H3 methylated at Lys4.
Includes This ChIPAb+ Monomethyl-Histone H3 (Lys4) -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Vertebrates
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.