Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Monomethyl-Histone H3 (Lys9)α, Mouse monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 17680MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-680-MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-680-MI Supplier MilliporeSigma™ Supplier No. 17680
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Specifically detects Monomethyl-Histone H3 (Lys9)α Clone: in Vertebrate samples, and it is validated for ChIP, ChIP-seq, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Monomethyl-Histone H3 (Lys9)α
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-monomethyl-Histone H3 (Lys9) (mouse monoclonal IgG2aқ, clone CMA306). One vial containing 50μg of protein G purified antibody in 50μL PBS containing 0.05% sodium azide. Normal Mouse IgG. Two vials containing 25μg purified Mouse IgG in 25μL storage buffer containing 0.1% sodium azide. ChIP Primers GAPDH Coding region. One vial containing 75μL of 5μM of each primer specific for a region of the human GAPDH coding region. FOR: GGC TCC CAC CTT TCT CAT CC; REV: GGC CAT CCA CAG TCT TCT GG
Gene Accession No. Q66I33
Host Species Mouse
Immunogen The monomethyl-histone H3 (Lys9) purified antibody is made against a synthetic peptide (monomethylated at Lys9) corresponding to amino acids 1-18 of Histone H3.
Purification Method Protein G purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NM_003493
Test Specificity Recognizes histone H3, Mr 17kDa, monomethylated at Lys9.
Includes This ChIPAb+ Validated Antibody and Primer Set conveniently includes the antibody, matched IgG negative control antibody and set control PCR primers that detect a known positive locus.
Target Species Vertebrates
Content And Storage −20°C for up to 12 months; Avoid repeated freeze/thaw cycles
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.