Learn More
MilliporeSigma™ Mouse iPS Reprogramming Boost Supplement
Catalog No. SCM087 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
SCM087 1 Kit
1 options
Catalog No. SCM087 Supplier MilliporeSigma™ Supplier No. SCM087
Add to Cart
Add to Cart
enhances the efficiency of mouse iPS colony formation by at least 3-fold.
Small molecules targeting specific signaling pathways have been shown to enhance the efficiency and/or replace transcription factors involved in somatic cell reprogramming. EMD Millipore has screened multiple chemical compounds and selected the compounds in this kit based on their effects on enhancement of reprogramming efficiency as assessed by the number and overall quality of mouse iPS colonies generated.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Kit contains sufficient quantities to supplement over 250mL of medium. Methylation Modulator-A Supplement (1000X): (Mfr. No. CS204419) One (1) vial containing 300μL of the inhibitor in water. GSK3β Inhibitor Supplement (1000X): (Mfr. No. CS204418) One (1) vial containing 300μL of the inhibitor in high quality DMSO. TGF-β RI Kinase Inhibitor II Supplement (1000X): (Mfr. No. CS204458) One (1) vial containing 300μL of the inhibitor in high quality DMSO. Store at -20°C
For Use With (Application) Enhancing efficiency of mouse iPS colony formation by at least 3-fold
Quantity 1 Kit