Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MQuant™ Aluminum Test Strips

Catalog No. M1100150001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1100150001 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1100150001 Supplier MilliporeSigma™ Supplier No. 1.10015.0001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Reagent

High-tech, fast and simple means for semiquantitative testing

Uses aurin tricarboxilic acid method

  • Small and easy to handle for on-site use
  • Fast results
  • Ideal for screening
  • PET film and low reagent content for easier disposal

  • Store cool and dry for use up to expiration date
  • Close tube immediately after removing strip to protect the remainder

  • Graduation: 10-25-50-100-250mg/L Al

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance 29 CFR 1910.1200
For Use With (Application) Used to check the integrity and efficiency of filtering systems as part of a general plan of in process controls
For Use With (Equipment) Filtering system
Graduations 10-25-50-100-250mg/L
Quantity 100 Tests
Storage Requirements 15°C to 25°C
Test Method Colorimetric
Type Aluminum Test Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.