Learn More
MilliporeSigma™ MQuant™ Formaldehyde Test Strips
Catalog No. M1100360001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
M1100360001 Each
1 options
Catalog No. M1100360001 Supplier MilliporeSigma™ Supplier No. 1.10036.0001
Add to Cart
Add to Cart
Includes
Reagent
High-tech, fast and simple means for semiquantitative testing
Uses triazole method
  • Small and easy to handle for on-site use
  • Fast results
  • Ideal for screening
  • PET film and low reagent content for easier disposal
  • Graduations: 10-20-40-60-100mg/L
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) For measuring formaldehyde, which is used as a disinfectant for surfaces and medical equipment, and also as a raw material for binders and fixing agents
Graduations 10-20-40-60-100mg/L
Includes 2 bottles Fo-1 reagent, 1 test vessel
Quantity Each
Storage Requirements 2°C to 8°C
Test Method Triazole
Type Formaldehyde Test Strip