Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MQuant™ Glucose Test Strips

Catalog No. M1178660001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1178660001 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1178660001 Supplier MilliporeSigma™ Supplier No. 1.17866.0001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-tech, fast and simple means for semiquantitative testing

Uses an enzymatic reaction to indicate concentration

  • For the determination of glucose in fruit jiuces and food
  • Store at 2° to 8°C
  • Small and easy to handle for on-site use
  • Fast results
  • Ideal for screening
  • PET film and low reagent content for easier disposal

  • Graduations: 10-25-50-100-250-500mg/L

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) For determining glucose in fruit juices and food
For Use With (Equipment) Colorimetric
Graduations 10-25-50-100-250-500mg/L
Quantity 50 Tests
Range 10 to 500
Storage Requirements 2°C to 8°C
Test Method Enzymatic reaction
Type Glucose Test Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.