Learn More
MilliporeSigma™ MQuant™ Lead Test Strips
Catalog No. M1100770001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
Catalog No. Quantity
M1100770001 100 Tests
1 options
Catalog No. M1100770001 Supplier MilliporeSigma™ Supplier No. 1.10077.0001
Add to Cart
Add to Cart
Includes
Reagent
High-tech, fast and simple means for semiquantitative testing
Uses the rhodizonic acid method.
  • Small and easy to handle for on-site use
  • Fast results
  • Ideal for screening
  • PET film and low reagent content for easier disposal
  • Graduations: 20-40-100-200-500mg/L
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) The lead test is suitable in environmental analysis, not only in all aqueous media but also on metal surfaces it can be used to distinguish very easy between plumbiferous and non-plumbiferous metals
Graduations 20-40-100-200-500mg/L
Includes 1 bottle of reagent Pb-1, 1 test vessel
Quantity 100 Tests
Range 20 to 500 mg/L
Storage Requirements 15°C to 25°C
Test Method Rhodizonic acid
Type MQuant™ Lead Test Strip