Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MQuant™ Potassium Iodide-Starch Reagent Papers

Catalog No. M1095120003 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
3 Rolls
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1095120003 3 Rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1095120003 Supplier MilliporeSigma™ Supplier No. 1.09512.0003
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Color scale

High-tech, fast and simple means for semiquantitative testing

Can easily be used anywhere for semiquantitative detection.

Used for the determination of oxidizing agents.
Grade: Reagent Pharmocopaeia Europe

Store at 15° to 25°C

Order Info

Comes as three rolls of 4.8m each

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Potassium iodine paper is used for the determination of oxidizing agents
Quantity 3 Rolls
Storage Requirements 15°C to 25°C
Type MQuant™ Potassium Iodide-Starch Reagent Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.