Learn More
MilliporeSigma™ MQuant™ Quaternary Ammonium Compounds Test Strips
Catalog No. M1179200001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
Catalog No. Quantity
M1179200001 100 Tests
1 options
Catalog No. M1179200001 Supplier MilliporeSigma™ Supplier No. 1.17920.0001
Add to Cart
Add to Cart
High-tech, fast and simple means for semiquantitative testing
Uses an indicator method.
Small and easy to handle for on-site use
Fast results
Ideal for screening
PET film and low reagent content for easier disposal
Graduations: 10-24-50-100-250-500 mg/L as Benzalkonium chloride
Order Info
No. of tests: 100
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) The test strip can be used to check quaternary ammonium compounds, which have microbicidal properties and are mainly used in surface cleaners
Graduations 10-25-50-100-250-500mg/L
Quantity 100 Tests
Range 10 to 500 mg/L
Storage Requirements 15°C to 25°C
Test Method Indicator
Type MQuant™ Quaternary Ammonium Compounds Test Strip