Learn More
MilliporeSigma™ MQuant™ Tin Test Strips
Catalog No. M1100280001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
M1100280001 Each
1 options
Catalog No. M1100280001 Supplier MilliporeSigma™ Supplier No. 1.10028.0001
Add to Cart
Add to Cart
Includes
Reagent
High-tech, fast and simple means for semiquantitative testing
Uses a toluene-3,4-dithiol
Small and easy to handle for on-site use
Fast results
Ideal for screening
PET film and low reagent content for easier disposal
Graduations: 10-25-50-100-200mg/L
Order Info
No. of tests: 50
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Used to test for tin and tin compounds in electroplating baths, reducing agents, stabilizers, catalysts, disinfectants, fungicides and mordants used in the textile industry and, more especially, in liquid foods
Graduations 10-25-50-100-200mg/L
Includes Sn-1 reagent bottle, 1 test vessel
Quantity Each
Storage Requirements 15°C to 25°C
Test Method Toluene-3,4-dithiol
Type Tin Test Strip