Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MultiScreen™ 96-Well Assay Plates

Catalog No. matrnps50 Shop All MilliporeSigma Products
Change view
Click to view available options
Color:
Clear
White
Product Type:
MultiScreen Acceptor Plate
MultiScreen Plate
MultiScreen Transport Receiver Filter Plate
MultiScreen-IP Filter Plate
MultiScreen-IP Filter Plate w/ Underdrain
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type
MATRNPS50 Clear MultiScreen Transport Receiver Filter Plate
MAIPN4550 Clear MultiScreen-IP Filter Plate w/ Underdrain
MPC4NTR10 Clear MultiScreen Plate
MSSACCEPT0R White MultiScreen Acceptor Plate
MAIPNTR10 Clear MultiScreen-IP Filter Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MATRNPS50 Supplier MilliporeSigma™ Supplier No. MATRNPS50
Only null left
Add to Cart
Add to Cart

Provide tools to perform high-throughput studies of ADME processes—the investigation of the adsorption, distribution, metabolism, and excretion of potential drugs

Versatile separation system combines a microplate with Millipore's membrane filtration to simplify and optimize drug discovery techniques, biochemical assays, and traditional sample preparation.

  • Use with a vacuum manifold, automated liquid-handling system, or microplate scintillation counter—entire assays can be performed on one plate
  • Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days
  • Also ensures quantitative filtrate transfer to a receiver plate placed in bottom of manifold

Choice of filter materials

  • For high protein binding applications, use plates with Immobilon™-P PVDF or mixed cellulose esters For high nucleic acid binding, use plates with mixed cellulose esters, glass fiber, or positively charged DEAE membranes For charged biomolecule separations, choose from positively charged DEAE or negatively charged phosphocellulose membranes
  • For low biomolecule binding, use plates with hydrophilic Durapore™ PVDF, Biopore™ PTFE, or polycarbonate membranes
  • For assays that require solvent resistance, use plates with hydrophilic PTFE membranes

Each microporous membrane interacts distinctly with different proteins or biomolecules; therefore, it may be necessary to test different membrane types for each assay.

†Includes lid, filter plate, and receiver plate.

‡Includes extended centrifugal cover, filter plate, and receiver plate.

††Consists of a 96-place holder and a 96-well receiver plate preloaded with Microcon YM-10 filter units.

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Array 8 x 12
Color Clear
Product Type MultiScreen Transport Receiver Filter Plate
Material Styrene
No. of Wells 96
Sterility Non-sterile
For Use With (Application) PAMPA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.