Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MultiScreen™ 96-Well Assay Plates for Cell-Based Assays

Catalog No. p-2363323 Shop All MilliporeSigma Products
Change view
Click to view available options
Pore Size:
3 μm
5 μm
8 μm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
MAMIC3S10 3 μm
MAMIC5S10 5 μm
MAMIC8S10 8 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MAMIC3S10 Supplier MilliporeSigma™ Supplier No. MAMIC3S10
Only null left
Add to Cart
Add to Cart

Offer formats that allow a wide range of functional cell-based and whole-organism assays to be performed

Ideal for cell viability and proliferation, migration, invasion, and chemotaxis assays. Versatile separation system combines a microplate with Millipore's membrane filtration to simplify and optimize drug discovery techniques, biochemical assays, and traditional sample preparation.

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Membrane Polycarbonate
Array 8 x 12
Pore Size 3 μm
Color Clear
Material Styrene
No. of Wells 96
Sterility Sterile
Surface Treatment Tissue Culture
Filtration Area 0.28 cm2
For Use With (Application) Migration, invasion and chemotaxis
Quantity 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.