Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MultiScreen™ 96-Well Assay Systems

Catalog No. p-4919744 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PSRP004R5 5/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PSRP004R5 Supplier MilliporeSigma™ Supplier No. PSRP004R5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Perform cell growth, feeding, and analysis in one plate

  • Patented 96-well device supports epithelial cell growth and differentiation of the Caco-2 and other cell lines
  • After the formation of a differentiated Caco-2 monolayer, the assembly can be used to measure the rate of drug transport across the Caco-2 cell barrier
  • All procedures are designed to be carried out in a single device
  • Apical and basolateral access ports allow contamination-free access to cells, easy feeding and media changes, and effective sample analysis
  • Increased sample throughput when compared to 24-well systems
  • Use manually or with automated systems
  • Versatile format with 96- or single-well feeder trays

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid With Lid
Membrane Polyethylene Terephthalate
Array 8 x 12
Pore Size 1 μm
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Sterile
Filtration Area 0.12 cm2
For Use With (Application) Cell attachment, growth and differentiation for many cell applications
Includes 96-well cell culture plate, single well feeder tray, 96-well receiver tray, lid
Quantity 5/Pk.
Dimensions (L x W x H) 0.5 x 3.33 x 1.03 in. (1.28 x 8.48 x 2.62 cm)
Length (Metric) 1.28 cm
Width (Metric) 8.48 cm
Height (Metric) 2.62 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.