Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MultiScreenHTS 384-Well Filter Plates

Catalog No. MZHVN0W50 Shop All MilliporeSigma Products
Change view
Click to view available options
Pore Size:
0.45 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
MZHVN0W50 0.45 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MZHVN0W50 Supplier MilliporeSigma™ Supplier No. MZHVN0W50
Only null left
Add to Cart
Add to Cart

Use in automated, high throughput applications, including biological assays and sample preparation. MultiScreenHTS 384-Well Filter Plates are non-sterile, single use plates available in several mesh materials and pore sizes to meet the needs of different applications.

For use in receptor-ligand binding, general sample preparation, enzyme assays with precipitation and DNA purification.
  • Rigid sidewalls allows full compatibility with automated/robotic systems
  • Unique plate design and individually sealed filter discs prevent cross-talk between wells
  • Opaque plastic lids prevents light transfer between wells during direct scintillation counting
  • Barcoding available
  • Meet ANSI™/SBS standards
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Membrane Durapore™ PVDF
Array 16 x 24
Pore Size 0.45 μm
Color Opaque
Material Styrene
No. of Wells 384
Sterility Non-sterile
Filtration Area 0.09 cm2
For Use With (Application) Receptor binding assays, protein kinase, enzyme assays
Quantity 50
Dimensions (L x W x H) 1.28 x 8.55 x 1.44 cm
Length (Metric) 1.28 cm
Width (Metric) 8.55 cm
Height (Metric) 1.44 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.