Learn More
MilliporeSigma™ Myeloperoxidase, Human Polymorphonuclear Leukocytes, Protein
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Formulation | Lyophilized from 100 to 700mM NaCl, 50mM sodium acetate buffer, pH 6.0. |
| Preparation Method | H2O (volume is lot-specific and can be found on the product label). Further dilute in 100mM NaCl, 50mM sodium acetate, pH 6.0. Enzyme may precipitate out of solution in low ionic strength buffers or in distilled H2O. |
| Purification Method | SDS-PAGE |
| Quantity | 100 μg |
| Immunogen | Myeloperoxidase |
| Storage Requirements | Following reconstitution, aliquot and freeze (−20°C or −70°C). Stock solutions are stable for up to 1 year at −20°C or −70°C. |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Native |
| Form | Lyophilized |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.