Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ N-CoR, Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710260MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-102-60MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-102-60MI Supplier MilliporeSigma™ Supplier No. 1710260
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects N-CoR Clone: in Human, Mouse samples, and it is validated for ChIP, Western Blotting

In the absence of ligands, the corepressor N-CoR represses nuclear hormone receptor-mediated gene transcription. In the presence of ligands, these hormone receptors activate gene expression and play critical roles in development, growth, and homeostasis. N-Cor-mediated transcriptional repression is facilitated through the formation of histone deacetylase (HDACs) N-CoR complexes or by conjugation from small ubiquitin-like modifier 1 (SUMO-1). It is shown that the SANT1 domain of N-CoR serves as a binding region for SUMO-E2 conjugating enzyme Ubc9 and SUMO-E3 ligase Pias1, which is necessary for N-CoR-mediated repression of respective reporter genes. SUMOylation-deficient N-COR knockout mice display a signification reduction in the repression of gene transcription and result in defects involving the differentiation and development of neural cells into erythrocytes and thymocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen N-CoR
Applications ChIP Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-N-CoR (Rabbit Polyclonal). One vial containing 75μg of affinity purified polyclonal in buffer containing 0.1M Tris-glycine (pH 7.4) and 150mM NaCl with 0.05% sodium azide before the addition of 30% glycerol. Concentration: 0.7mg/mL. Normal Rabbit IgG One vial containing 125μg of Rabbit IgG in 125μL of storage buffer containing 0.05% sodium azide. ChIP Primers, p21. One vial containing 75μL of each primer (5μM) specific for the human p21 (WAF1/CIP1/CDKN1A) promoter. FOR: CCC ACA GCA GAG GAG AAA GAA; REV: CTG GAA ATC TCT GCC CAG ACA
Gene Accession No. Q60974.
Gene Symbols Ncor1, Rxrip13
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a region between amino acids 1510 and 1540 of the N-CoR protein.
Purification Method Affinity Purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_035438
Includes This ChIPAb+ N-CoR -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Mouse
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.