Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Nanog Mouse monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 17655MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-655-MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-655-MI Supplier MilliporeSigma™ Supplier No. 17655
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Specifically detects Nanog Clone: in Human samples, and it is validated for ChIP, Immunocytochemistry, Western Blotting

NANOG (Homeobox protein NANOG) is a member of the Nanog homeobox family of DNA-binding proteins. It is expressed in embryonic stem cells and confers pluripotency on these cells. Once embryonic stem cells become differentiated, NANOG expression is suppressed.  NANOG is involved in the Hedgehog/Gli1 signaling pathway which has been implicated in the development and growth of various types of tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nanog
Applications ChIP Assay, Immunocytochemistry, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-Nanog (Mouse Monoclonal). One vial containing 110μL (0.7 mg/mL) purified mouse monoclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4),150mM NaCl, and 0.05% sodium azide, before the addition of 30% glycerol. Normal Mouse IgG. One vial containing 125μg of purified mouse IgG in 125μL storage buffer containing 0.1% sodium azide. ChIP Primers, miR-302. One vial containing 75μL of 5μM of each primer specific for human miR-302 cluster promoter region (chr4:113570181-113570261, hg19 build). FOR: CCC GTG GAA GCA ATC TAT TTA TTT; REV: TGT GTT TCT ATC TGG AGG AAC TCT GT
Gene Accession No. Q9H9S0
Gene Symbols NANOG
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human NANOG.
Purification Method Protein G purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_079141
Includes Use ChIPAb+ Nanog ChIP Validated mouse Monoclonal for the detection of Nanog.
Target Species Human
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.