Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Ndiff™ Chemicon™ Neuro-27 Medium Supplement (100x)

Catalog No. SCM013 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCM013 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCM013 Supplier MilliporeSigma™ Supplier No. SCM013
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

a serum-free supplement containing antioxidants and other factors that has been developed for the in vitro propagation and maintenance of undifferentiated murine embryonic stem (ES) cells in serum free medium.

NDiff™ Neuro-27 Medium Supplement is a serum-free supplement containing antioxidants and other factors that has been specifically developed for the in vitro propagation and maintenance of undifferentiated murine embryonic stem (ES) cells in serum free medium (Nichols J, 2006). This product can also be used to differentiate murine ES cells into post-mitotic neurons particularly via monolayer differentiation (Ying QL, 2003).
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Serum Level Serum-free
For Use With (Application) Stem cell culture and cell culture
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.