Learn More
MilliporeSigma™ Neurogenesis Kit
Catalog No. SCR603 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
SCR603 1 Kit
1 options
Catalog No. SCR603 Supplier MilliporeSigma™ Supplier No. SCR603
Add to Cart
Add to Cart
a robust and user-friendly kit that contains the necessary serum free/chemically defined media and reagents for the rapid production of expandable neural progenitor cells and terminally differentiated neurons from pluripotent human ES/iPS cells
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Cell Differentiation, Neuroscience
Quantity 1 Kit
Content And Storage 1) Human ES/iPS Neural Induction Medium (Mfr. No. SCM110): 125mL, Human ES/iPS Neuronal Differentiation Medium (Mfr.. No. SCM111): 100mL, 2) ENStem-A Neural Expansion Medium (Mfr. No. SCM004): 500mL. Store at -20°C.