Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Neuropeptide Y, Human, Calbiochem

Catalog No. 052320051MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-232-0051MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-232-0051MG Supplier MilliporeSigma™ Supplier No. 052320051MG
Add to Cart
Add to Cart
A potent vasoconstrictor. Reversibly inhibits Ca2+-activated K+ channels in vascular smooth muscle cells.
TRUSTED_SUSTAINABILITY

Specifications

Species Human
Purification Method HPLC
Purity ≥97%
CAS 90880-35-6
Color White to Off-white
Content And Storage Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 3 months at −20°C.
Format Lyophilized Solid
Formulation Supplied as trifluoroacetate salt. Sold on the basis of peptide content.
Product Type Neuropeptide Y
Quantity 1 mg
Regulatory Status RUO
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.