Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ BacVector™ Insect Cell Medium
GSA_VA

Catalog No. 705903 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-590-3 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-590-3 Supplier MilliporeSigma™ Supplier No. 705903
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Optimized for growth of Sf9 Insect Cells

  • Serum-free medium
  • Recommended for cotransfection of BacVector Triple Cut Virus DNA or BacMagic™ DNA
  • Transfer plasmids to construct recombinant baculoviruses
  • Compatible with Insect GeneJuice™ Transfection Reagent
  • Recommended for transient expression from pIEx™ and pBiEx™ vector recombinants, plaque assays using agarose overlay technique

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Insect Cell Medium
Quantity 1 L
Content And Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.