Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ OsteoMAX-XF™ Differentiation Medium

Catalog No. SCM121 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCM121 100 mL/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCM121 Supplier MilliporeSigma™ Supplier No. SCM121
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

specially formulated to readily differentiate human mesenchymal stem cells (MSCs) into mature osteocytes as assessed by alizarin red and alkaline phosphatase staining.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Stem Cell Culture, Cell Culture
Quantity 100 mL/Kit
Content And Storage -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.