Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ P2X7 Receptor (576-595), Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 5061071SET Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
50-610-71SET 1 Set Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-610-71SET Supplier MilliporeSigma™ Supplier No. 5061071SET
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects P2X7 Receptor (576-595) Clone: in Rat samples, and it is validated for Immuno Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen P2X7 Receptor (576-595)
Applications Immunoblot
Classification Polyclonal
Conjugate Unconjugated
Formulation 50μl antibody lyophilized from PBS, 1% BSA, pH 7.4 and 40μg lyophilized control peptide (immunogen).
Host Species Rabbit
Immunogen a synthetic peptide [(C)KIRKEFPKTQGQYSGFKYPY] corresponding to amino acids 576-595 of rat P2X7 receptor, conjugated to receptor protein
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the P2X7 receptor protein. Supplied with a control peptide.
Includes 50μL antibody lyophilized from PBS, 1% BSA, pH 7.4 and 40μg lyophilized control peptide (immunogen).
Target Species Rat
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.