Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ p53α, Mouse monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 17613MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-613-MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-613-MI Supplier MilliporeSigma™ Supplier No. 17613
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects p53α Clone: in Human samples, and it is validated for ChIP, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen p53α
Applications ChIP Assay, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-p53 (mouse monoclonal IgG). One vial containing 25μg of of protein A purified mouse IgG2a in 25μL of storage buffer (PBS, pH 7.4, containing 0.05% sodium azide and 1mg/mL BSA). Normal Mouse IgG. One vial containing 25μg of normal mouse IgG in 25μL volume. ChIP Primers p21. One vial containing 75μL of 5μM of each primer specific for a region of the human p21 (WAF1/CIP1/CDKN1A) promoter. FOR: GTG GCT CTG ATT GGC TTT CTG; REV: CTG AAA ACA GGC AGC CCA AG
Gene Accession No. P04637
Gene Symbols P53, TP53
Host Species Mouse
Purification Method Protein A purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_000537
Test Specificity Recognizes p53 in wild type and mutant forms of human origin.
Includes This ChIPAb+ p53 -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human
Content And Storage −20°C for one year from date of shipment
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.