Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ p53 Sampler kit

Catalog No. ASK071EA Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ASK071EA 1 Kit Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ASK071EA Supplier MilliporeSigma™ Supplier No. ASK071EA

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Specifically detects p53 Clone: in samples, and it is validated for

The human p53 tumor suppressor gene encodes a 393 amino acid phosphoprotein that exhibits sequence-specific DNA binding and directly interacts with various cellular and viral proteins. p53 is the most commonly mutated gene in human cancer, with the majority of the mutations being amino acid substitutions. The normal function of p53 is to effect cell cycle arrest at the G1 and G2 checkpoints in response to DNA damage. This checkpoint function is executed by accumulation of p53 followed by induction of the GADD45, WAF1 and MDM2 genes. The current model of p53 function postulates that p53 senses DNA damage and arrests the cell cycle in either the G1 or G2 phases to allow DNA repair to take place. If repair is not successful, p53 initiates programmed cell death, thus preventing the propagation of genetic defects to successive generations of cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p53
Classification Monoclonal
Conjugate Unconjugated
Formulation 20μg of each antibody in 200μL (100μg/ml) of 0.05 M sodium phosphate buffer containing 0.1% sodium azide and 0.2% gelatin.
Host Species Mouse
Quantity 1 Kit
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity 20μg of various p53 antibodies.
Includes 20μg of each antibody in 200μL (100μg/mL) of 0.05M sodium phosphate buffer containing 0.1% sodium azide and 0.2% gelatin.
Content And Storage 2°C to 8°C
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.