Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Packs and Accessories for Milli-RO™ Compact Water Systems: RO Cartridges

Catalog No. CDRCH12HG Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With (Equipment):
Elix™, RiOs™ 3, RiOs, Milli-RO™
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
CDRCH12HG Elix™, RiOs™ 3, RiOs, Milli-RO™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDRCH12HG Supplier MilliporeSigma™ Supplier No. CDRCH12HG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RO cartridges for Milli-RO Compact Water Systems.

High-recovery reverse osmosis cartridge removes inorganic ions.

TRUSTED_SUSTAINABILITY

Specifications

Purification Method Reverse osmosis
For Use With (Equipment) Elix™, RiOs™ 3, RiOs, Milli-RO™
Model Hi-flux RO
Type Cartridge
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.