Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Pam3Cys-Ser-(Lys)4, Hydrochloride, Protein

Catalog No. 5063502MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-635-02MG 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-635-02MG Supplier MilliporeSigma™ Supplier No. 5063502MG
Add to Cart
Add to Cart
A cell-permeable, water-soluble synthetic cationic lipohexapeptide analog of the immunologically active N-terminal portion of bacterial lipoprotein that potently activates monocytes and macrophages. Induces the release of IL-1, IL-6, TNF-α, superoxide, and NO. Promotes the translocation of NF-κB, enhances tyrosine protein phosphorylation and activation of ERK1/2 and MEK1/2. Acts as a selective agonist of TLR2 (Toll-like receptor 2).
TRUSTED_SUSTAINABILITY

Specifications

Preparation Method H2O (2mg/mL)
Purification Method HPLC
Quantity 2 mg
Immunogen Pam3Cys-Ser-(Lys)4, Hydrochloride
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Avoid freeze/thaw cycles of solutions.
Regulatory Status RUO
Recombinant Native
Form Lyophilized
Purity or Quality Grade ≥90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.