Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Parathyroid Hormone 1-34, Human, Calbiochem

Catalog No. 052355011MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-235-5011MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-235-5011MG Supplier MilliporeSigma™ Supplier No. 052355011MG
N-terminal active peptide fragment of the native hormone that functions as a regulatory factor in the homeostatic control of Ca2+ and phosphate metabolism. Regulates Ca2+ and phosphate flux across cellular membranes in bone and kidney, resulting in increased serum Ca2+ levels. Increases the activity of osteoblasts and osteoclasts. Increases tubular reabsorption of Ca2+ and stimulates renal hydroxylation of 25-(OH)-Vitamin D3 to 1,25-(OH)2-Vitamin D3. Increases the production of inhibitory IGF binding protein 4 in bone cells.
TRUSTED_SUSTAINABILITY

Specifications

Species Human
Purification Method HPLC
Purity ≥95%
CAS 52232-67-4
Color White to Off-white
Content And Storage Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 3 months at −20°C.
Format Lyophilized Solid
Formulation Supplied as an acetate salt.
Product Type Parathyroid Hormone 1-34
Quantity 1 mg
Regulatory Status RUO
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.