Learn More
MilliporeSigma™ pBAC-3 DNA
Catalog No. 700883 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
70-088-3 10 μg
1 options
Catalog No. 70-088-3 Supplier MilliporeSigma™ Supplier No. 700883
Add to Cart
Add to Cart
pBAC™-3 is a baculovirus transfer plasmid compatible with BacVector™-1000, -2000, or -3000 Triple Cut Virus DNA for low background transfection and efficient utilization of the polh promoter and expression of secreted proteins. A 20 amino acid gp64 signal peptide sequence is capable of directing high levels of protein into the secretory pathway of infected insect cells.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Quantity 10 μg