Learn More
MilliporeSigma™ pBAC 4x-1 DNA
Catalog No. 700453 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
70-045-3 10 μg
1 options
Catalog No. 70-045-3 Supplier MilliporeSigma™ Supplier No. 700453
Add to Cart
Add to Cart
pBAC4x-1 is a baculovirus transfer plasmid designed for cloning and co-expression of up to four target genes or multiple copies of the same gene in insect cells. The plasmid contains two polh promoters and two p10 promoters, each of which is upstream of unique cloning sites for sequential insertion of target genes.
TRUSTED_SUSTAINABILITY
Specifications
Promoter Polyhedrin (2 copies), p10 (2 copies)
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Quantity 10 μg