Learn More
MilliporeSigma™ pBiEx-1 DNA Vector
Catalog No. 712343 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 μg
Catalog No. Quantity
71-234-3 20 μg
1 options
Catalog No. 71-234-3 Supplier MilliporeSigma™ Supplier No. 712343
Add to Cart
Add to Cart
The pBiEx™ vectors are designed for cloning and high-level expression of proteins in both bacteria and in transiently transfected Spodoptera-derived insect cells. In E. coli, target gene transcription is regulated by the tightly controlled T7lac promoter, and a ribosome binding site (rbs) directs translation initiation. In insect cells, transcription is driven by the AcNPV-derived hr5 enhancer and immediate early promoter, IE1.
TRUSTED_SUSTAINABILITY
Specifications
Promoter T7lac
Content And Storage Store at -20°C.
Quantity 20 μg
For Use With (Application) Cloning and high-level expression of proteins in both bacteria and in transiently transfected Spodoptera-derived insect cells