Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ PCAF, Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710532MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-105-32MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-105-32MI Supplier MilliporeSigma™ Supplier No. 1710532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects PCAF Clone: in Human samples, and it is validated for ChIP, Western Blotting

P/CAF(sometimes called KAT2B) is a transcription cofactor with a histone acetyltransferase (HAT) activity. Consistent with this protein playing a direct role in transcriptional regulation, P/CAF can acetylate core histones H3 and H4, chromatin proteins HMG17 and HMG I(Y), as well as human immunodeficiency virus Tat, transcription factor MyoD, and general transcription factors TFIIE and TFIIF. A role for P/CAF in cell growth and differentiation is suggested by its interaction in vivo and in vitro with p300/CBP, two large nuclear proteins that bind to sequence-specific factors involved in cell growth/differentiation. A role for P/CAF in cell division is also suggested by data showing that this protein competes with E1A for binding sites in p300/CBP to counteract E1A's mitogenic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCAF
Applications ChIP Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-PCAF (Rabbit Polyclonal). One vial containing 17.5μg purified rabbit polyclonal in 25μL buffer containing Tris-citrate/phosphate buffer (pH 7 to 8) and 0.09% sodium azide, before the addition of 30% glycerol (0.7mg/mL final). Normal Rabbit IgG. One vial containing 125μg of purified Rabbit IgG in 125μL storage buffer containing 0.05% sodium azide. ChIP Primers, PRAME. One vial containing 75μL of 5μM of each primer specific for human preferentially expressed antigen in melanoma (chr22:22901678+22901804, hg19 build). FOR: GAA GCC TCG CCG GAA CTC; REV: TCC TCC CCA TCT CTG CAG AA
Gene Accession No. Q92831
Gene Symbols GCN5, GCN5L, P/CAF, P, CAF, PCAF, KAT2B
Host Species Rabbit
Immunogen Recombinant protein corresponding to human PCAF (a.a. 782-832).
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_003875
Test Specificity Recognizes PCAF in regions between a.a. 782 to 832.
Includes This ChIPAb+ Validated Antibody and Primer Set conveniently includes the antibody, matched IgG negative control antibody and set control PCR primers that detect a known positive locus.
Target Species Human
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.