Learn More
MilliporeSigma™ pCDF-1b DNA Vector
Catalog No. 713303 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
71-330-3 10 μg
1 options
Catalog No. 71-330-3 Supplier MilliporeSigma™ Supplier No. 713303
Add to Cart
Add to Cart
The pCDF-1b plasmid features coexpression capabilities as well as the ability to express fusion proteins with a N-terminal His·Tag that results in native protein after purification and cleavage. This plasmid carries an origin derived from CloDF13 (1) and streptomycin/spectinomycin resistance, allowing for the option of coexpression with many other Novagen T7-based expression vectors. The vector contains a T7 promoter, lac operator, ribosome binding site (rbs), an amino terminal His·Tag™ coding sequence, and multiple cloning site (MCS) regions designed to allow the generation of target proteins with minimal vector-encoded fusion.
TRUSTED_SUSTAINABILITY
Specifications
Promoter T7
Content And Storage Store at or below -70°C.
Quantity 10 μg