Learn More
MilliporeSigma™ pCDF-2 Ek/LIC Vector Kit
Catalog No. 713373 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 Reactions
Catalog No. Quantity
71-337-3 20 Reactions
1 options
Catalog No. 71-337-3 Supplier MilliporeSigma™ Supplier No. 713373
Add to Cart
Add to Cart
The pCDF-2 Ek/LIC Vector is a LIC-compatible derivative of pCDF-1b. The plasmid contains a strong T7lac promoter and an amino terminal His·Tag™ coding sequence immediately followed by an Ek/LIC cloning site designed to allow the generation of fusion proteins with minimal vector-encoded sequence. An optional C-terminal S·Tag™ coding sequence is compatible with purification, detection, and quantification.
TRUSTED_SUSTAINABILITY
Specifications
Promoter T7lac
Content And Storage Store at or below -70°C.
Quantity 20 Reactions