Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ pCOLADuet-1 DNA Vector

Catalog No. 714063 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-406-3 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-406-3 Supplier MilliporeSigma™ Supplier No. 714063
Add to Cart
Add to Cart
pCOLADuet™-1 is designed for the coexpression of two target genes from a single plasmid. The vector encodes two multiple cloning sites (MCS) each of which is preceded by a T7 promoter, lac operator, and ribosome binding site (rbs). MCS-1 encodes the six-amino acid His·Tag™ sequence for the creation of a N-terminal fusion and MCS2 encodes the 15 amino acid S·Tag™ peptide after the last restriction site for the creation of a C-terminal fusion if desired.
TRUSTED_SUSTAINABILITY

Specifications

Marker Gene Kanamycin resistance
Promoter T7
Content And Storage Store at -20°C.
Quantity 10 μg
For Use With (Application) Coexpression of two target genes from a single plasmid

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.